Z743744
ART® Low Retention SoftFit L Reload System
HLT 1000L, size 1000 μL, hinged rack, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
feature
HLT 1000L
hinged rack
packaging
pkg of 4X768 ea
manufacturer/tradename
Molecular BioProducts 3782-05-HR
technique(s)
PCR: suitable
size
1000 μL
Looking for similar products? Visit Product Comparison Guide
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service