Key Documents
Z743206
Julabo® bath accessories
assembly cooling coil, for CORIO C and CD ranges, pack of 1 ea
Select a Size
1 EA
R$1,433.00
R$1,433.00
Estimated to ship onMay 21, 2025
Select a Size
Change View
1 EA
R$1,433.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
R$1,433.00
Estimated to ship onMay 21, 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970100
General description
Built-in cooling coil (9mm outer diameter) for counter-cooling with cooling water, can be used for tempering tasks close to the ambient temperature. A hose with an inner diameter of 10/12 mm is required. For use with Julabo® baths/circulators.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)