Key Documents
Z743163
Julabo® bath covers
flat cover to fit CORIO C-B13/17 and CD-B13 /17
Select a Size
1 EA
R$2,974.00
R$2,974.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$2,974.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
R$2,974.00
Check Cart for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970290
General description
Stainless Steel Flat bath cover for 13L and 17L bath tanks. The use of a bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath. Suitable for use with Julabo® CORIO C-B13, C-B17, CD-B13, CD-B17.
Features and Benefits
  • reduces evaporation
  • prevents contamination of samples
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)