This item has a low temperature range from -20 °C to -40 °C with an upper limit of 150 °C. The temperature accuracy (stability) is ±0.03 °C.
Z743065
Julabo® CORIO™ CD refrigerated/heating circulator
CD-300F, bath useable volume 3 L (4 L), temp -25 °C, accuracy: 150 °C, AC/DC input 230 V AC, Schuko plug
Select a Size
R$37,184.00
Select a Size
About This Item
R$37,184.00
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9012703
temp measuring range
-25 °C, accuracy: 150 °C
bath useable volume
3 L (4 L)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy change-over from internal to external circulation and vice versa
- External pump connections (M16x1)USB interface
- Space-saving cooling coil design provides more usable space in the bath tank
- Bath lid and drain tap included
- Removable ventilation grid
- Refrigeration unit without side vents
- Class III (FL) according to DIN 12876-1
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the temperature range of this bath?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service