Z743026
Julabo® CORIO™ CD heating circulator with open bath
CD-B39, capacity 35 L (41 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
Select a Size
R$28,170.00
Select a Size
About This Item
R$28,170.00
Recommended Products
material
stainless steel bath
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9012439
temperature measuring range
20-150 °C
capacity
35 L (41 L)
external W × L × H
54 cm × 34 cm × 57 cm
opening W × L × H
33 cm × 30 cm × 30 cm
General description
Your advantages
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- USB connection
- High-quality bath tanks made of stainless steel
- Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)