Documents
Z742867
Kern PLS series precision balances
Kern PLS 420-3F, weighing capacity 420 g, accuracy: 0.001 g, precision: ±0.004 g
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
AC/DC input
110 - 240 V AC, universal plug set
packaging
pack of 1 ea
manufacturer/tradename
Kern PLS 420-3F
weighing capacity measuring range
420 g, accuracy: 0.001 g, precision: ±0.004 g
Related Categories
General description
Range of precision balances with CAL adjusting program for quick setting of the balance accuracy.
Ergonomically optimised keypad for left and righthanded users