Key Documents
Z742839
Kern PCB series precision balances, rechargable battery pack
pack of 1 ea
Select a Size
1 EA
R$1,025.00
R$1,025.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$1,025.00
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
R$1,025.00
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Kern PCB-A01
General description
Internal rechargeable NiMH battery pack PCB-A01, with Euro, UK, US, CH plug types. Operating time up to 48 h, charging time approx. 8 h, Auto-Off function preserves the rechargeable battery pack. Suitable for models from the KERN PCB, KERN ECB, KERN ECE and KERN FKB-A series.
Choose from one of the most recent versions:
Certificates of Analysis (COA)