Key Documents
Z742265
DURAN® GL 56 Bottle Tag
blue silicone
Synonym(s):
292435659
Select a Size
20 EA
R$830.00
R$830.00
Check Cart for Availability
Select a Size
Change View
20 EA
R$830.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$830.00
Check Cart for Availability
material
blue silicone
packaging
pack of 20 ea
manufacturer/tradename
DURAN® 292435659
General description
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Features and Benefits
  • Manufactured from silicone rubber
  • The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
  • The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and reusable
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)