Z740916
Aldrich® Essentials graduated cylinder, class B, meets DIN ISO 4788
capacity 500 mL, subdivision, 5.0 mL, tol. 5.0 mL, Graduated "To Contain", plastic bumper, hex base
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
subdivision 5.0 mL
packaging
pkg of 1 ea
tol. measuring range
5.0 mL
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Clear borosilicate glass graduated cylinders with safety hexagonal base. Pour spout is offset of base point so that if cylinder tips the spout won′t be damaged. Hex base prevents the cylinder from rolling off a bench. Meets DIN ISO 4788 Class B tolerances and specifications. Complete with plastic bumper. Graduated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service