Skip to Content
Merck
All Photos(1)

Key Documents

Z699128

Enterococcus Broth

pack of 50 × 2 mL

Synonym(s):

agar broth, broth and agar

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
R$712.00

R$712.00


Estimated to ship onJune 01, 2025



Select a Size

Change View
50 EA
R$712.00

About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

R$712.00


Estimated to ship onJune 01, 2025


packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496120

application(s)

food and beverages

suitability

Enterococcus spp.

General description

Enterococcus Broth is a modified version of the improved media described by Slanetz and Bartley with TTC. The membrane filtration method is simple to perform, does not require confirmation and permits a direct count of enterococci in 48 hours.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service