packaging
pkg of 10 ea
manufacturer/tradename
3M
Related Categories
General description
3M high-performance filter material combines effective filtration with low breathing resistance, offering protection against fine dusts and oil/water based mists.
Approvals
- Unique 3 panel design fits a wide range of face shapes and sizes.
- 3M Cool Flow® valve removes heat, exhaled air and moisture which reduces the risk of misting eyewear.
- Blue straps denote 3M FFP2.
- Dolomite test "D" resists clogging for longer.
Approvals
- EN 149:2001
- CE-marked
- Category FFP2, Assigned Protection Factor 10 (NFP)
Legal Information
3M is a trademark of 3M Company
Cool Flow is a registered trademark of 3M Corp.