Skip to Content
Merck
All Photos(1)

Key Documents

Z685631

Tygon® formula 2375 laboratory tubing

I.D. × O.D. 4.8 mm × 8.0 mm, L 15 m (50 ft)

Synonym(s):

chemical resistant tubing, laboratory tubing, non-leaching tubing

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
R$1,080.00

R$1,080.00


Check Cart for Availability


Select a Size

Change View
1 PKG
R$1,080.00

About This Item

UNSPSC Code:
41105108
NACRES:
NB.51

R$1,080.00


Check Cart for Availability

packaging

pack of 50 feet

manufacturer/tradename

St. Gobain AJK00012

I.D. × O.D.

4.8 mm × 8.0 mm

L

15 m (50 ft)

wall thickness

1.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

Tygon laboratory tubing, formula 2375, is chemically resistant tubing. Clear, flexible tubing suitable for specialty chemical production and hazardous material handling.

  • Resistant to MEK, acids, bases, ketones, and alcohols
  • Plasticizer free
  • No contamination from leaching
  • Biocompatible
  • No disposal concerns
  • Releases only carbon dioxide and water upon incineration
  • Sterilize by gas or radiation

Specifications

Technical data:
Tensile Strength: 1900 psi (13.1 MPa)
Ultimate Elongation: 850%
Tensile Stress @ 100% elongation: 425 psi (2.9 MPa)
Tensile Set (@ 75% of ultimate elongation): 300%
Tear Resistance: 240 lb-f/inch (42 kN/m)
Compression Set Constant Deflection: 100% @ 158°F (70°C) for 22 hours
Brittle Temperature: < -75°F (<-59°C)
Low Temp. Flexibility: -103°F (-75°C)
Specific Gravity: 0.9
Water Absorption: 0.04% 24 hrs at 73°F (23°C)
Maximum Recommended Operating Temperature: 130°F
Color: Clear

Legal Information

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert Morhard et al.
IEEE transactions on bio-medical engineering, 67(8), 2337-2348 (2019-12-17)
Ethanol ablation, the injection of ethanol to induce necrosis, was originally used to treat hepatocellular carcinoma, with survival rates comparable to surgery. However, efficacy is limited due to leakage into surrounding tissue. To reduce leakage, we previously reported incorporating ethyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service