Z683256
J.Young high-vacuum PTFE valve, right angle
stopcock size 15 mm, side arm O.D. 19 mm
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
J. Young
side arm O.D.
19 mm
stopcock size
15 mm
General description
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
Features:
- PTFE piston with unique PTFE-sheathed elastomer O-ring seals
- Grease-free glass-to-PTFE seals
- High-vacuum to 10-6 torr