Z682039
Scaffdex CellVessel™ insert scaffolds
multifilament copolymer fiber scaffold, diam. 2.4 mm, sterile
Synonym(s):
3D scaffolds, cell scaffolds
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
sterility
sterile
manufacturer/tradename
Scaffdex V00002
diam.
2.4 mm
fiber L
100 mm
color
white
Looking for similar products? Visit Product Comparison Guide
General description
CellVessel is made of bioabsorbable multifilament (8-filaments) poly-96L/4D-lactide copolymer fiber. Scaffdex′s braided scaffold is made for research purposes, only and it should not be used in any kind of clinical settings. Poly-96L/4D-lactide copolymer has been tested with various cell types and it works especially well with fibroblasts.
CellVessel scaffolds are provided as gamma sterilized and with three widths. The shape of the cross-section of CellVessel varies a little bit from complete round to oval, depending on the size. If further sterilization is needed before cell culturing, ethylene oxide or ethanol rinse to avoid further decreasing of inherent viscosity and strenth properties is suggested.
Note: Should CellVessels be cut into wanted lengths by the use, a light heat treatment of the ends of the two bigger sizes should be done to avoid the unloading of the structure during testing.
CellVessel scaffolds are provided as gamma sterilized and with three widths. The shape of the cross-section of CellVessel varies a little bit from complete round to oval, depending on the size. If further sterilization is needed before cell culturing, ethylene oxide or ethanol rinse to avoid further decreasing of inherent viscosity and strenth properties is suggested.
Note: Should CellVessels be cut into wanted lengths by the use, a light heat treatment of the ends of the two bigger sizes should be done to avoid the unloading of the structure during testing.
Legal Information
CellVessel is a trademark of Scaffdex Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service