Z677647
Butoject™ butyl gloves
size M
Synonym(s):
butyl rubber gloves, chemical resistant gloves
Select a Size
R$617.00
Estimated to ship onMarch 24, 2025
Select a Size
About This Item
R$617.00
Estimated to ship onMarch 24, 2025
Recommended Products
material
black butyl rubber
black gloves
size
M
sterility
non-sterile
feature
CE compliant
latex free
powder-free: no
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
pack of 1 pair
manufacturer/tradename
KCL 897
L × thickness
360 mm × 0.3 mm
General description
Butoject is very comfortable to wear and allows good dexterity. To improve the grip on smooth and wet parts, this glove has an additionally roughened surface.
Features and Benefits
- Provides excellentprotection against aggressive chemicals, helping lab technicians work safely with harsh substances.
- Ensures wearing comfort and dexterity due to its high elasticity and ergonomic design, reducing fatigue during long tasks.
- Improved grip on wetsurfaces with a roughened surface, enhancing safety and handling of equipment.
- Made of durable butyl rubber, offering good mechanical resistance and extended usability, reducing the need for frequent replacement.
- Standard type EN 374 (cat.III) and EN 388 (cat. III) compliant, ensuring a high level of chemical and mechanical protection.
- Powder-free design minimizes contamination and is CE compliant for safe use in laboratory environments.
- Reusable, making it cost-effective for labs with frequent glove usage.
- 360 mm length and 0.3 mmthickness provide extensive coverage while maintaining flexibility and coldresistance.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)