Z676314
Vacuubrand chemistry vacuum system
AC/DC input 220 V AC, European 2-pin plug, Vacuubrand MD 4C NT+AK+EK
Synonym(s):
Vacuubrand 20736403, diaphragm pump
About This Item
Recommended Products
feature
CE compliant
(Number of heads / stages 4/3)
packaging
pack of 1 ea
manufacturer/tradename
Vacuubrand MD 4C NT+AK+EK
parameter
1.5 mbar vacuum
3 mbar vacuum (with gas ballast)
3.4/3.8 m3/h pumping speed
L × W × H
326 mm × 243 mm × 402 mm
inlet and outlet size
10 mm , connection nozzle
size
6-8 mm , coolant connection 2 x hose nozzle
Looking for similar products? Visit Product Comparison Guide
General description
The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The waste vapor condenser at the outlet (EK) is highly efficient and compact. The condenser enables near-100-percent solvent recovery, efficient recycling of solvents and active protection of the environment. Items supplied completely mounted, ready for use, with manual.
- Outstanding chemical resistance and superior vapor tolerance
- Exceptionally high performance even at low vacuum
- Excellent ultimate vacuum even with gas ballast
- Whisper quiet and very low vibration
- Excellent environmental friendliness due to efficient solvent recovery
- IP40 rated protection
Order 10 mm connecting tubing separately.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service