Documents
Z657212
IKA® overhead stirrer accessories
R 2723 telescopic stand
About This Item
UNSPSC Code:
41122800
NACRES:
NB.72
packaging
pack of 1 ea
manufacturer/tradename
IKA 0001412100
General description
Stable with H-shape base which prevents the stand from tipping backwards. Provides optimum stability required for larger, heavier instruments and attachments. The stand has an adjustment screw which can be compensated for an uneven laboratory table surface. Additionally equipped with a pneumatic spring stand rod, which enables heavy instruments/attachments to be raised and lowered smoothly and without difficulty.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG