Key Documents
Z657182
IKA® A11 basic grinding chambers
A 11.5 grinding chamber, 1/cs
Synonym(s):
grinders, grinding chambers, grinding mills
Select a Size
1 EA
R$3,730.00
R$3,730.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$3,730.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.74
R$3,730.00
Check Cart for Availability
feature
CE compliant
CSA approved
packaging
pack of 1
manufacturer/tradename
IKA 0002983100
size
80 mL , effective volume
General description
Replacement grinding bowl for use with the IKA A11 Basic Analytical Mill. Made of Tefcel (ETFE, glass fiber reinforced) with stainless steel inlet (AISI 316L), usable volume 80ml. Has good chemical and low temperature resistance down to -200 °C.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)