Key Documents
Z609633
Bohlender screw caps
GL 14 thread
Select a Size
10 EA
R$287.00
R$287.00
Check Cart for Availability
Select a Size
Change View
10 EA
R$287.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$287.00
Check Cart for Availability
manufacturer/tradename
Bohlender
availability
not available in USA and Canada
fitting
GL 14 thread
General description
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
Legal Information
Bohlender is a trademark of Bohlender GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)