Key Documents
Z607002
Bohlender PTFE beaker
capacity 25 mL, white
Select a Size
1 EA
R$190.00
R$190.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$190.00
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
R$190.00
Check Cart for Availability
material
PTFE
manufacturer/tradename
Bohlender
availability
not available in USA and Canada
capacity
25 mL
color
white
General description
With spout and reinforced rim, thick-walled, smooth interior surface.
Other Notes
Browse all plastic beakers
Legal Information
Bohlender is a trademark of Bohlender GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)