Documents
Z562564
Continuous flow cell for 130W ultrasonic processors
designed primarily for dispersing and homogenizing at rates up to 15 liters/hour
About This Item
UNSPSC Code:
41101500
NACRES:
NB.74
feature
autoclavable
packaging
pkg of 1 ea
manufacturer/tradename
Sonics & Materials 630-0566
Related Categories
General description
The continuous flow cell screws into the converter in place of the probe. Recommended only for the treatment of low viscosity samples which do not require extended
exposure to ultrasonics.
Designed primarily for dispersing and homogenizing at rates up to 15 liters/hour.
The vessel is fabricated from glass. The probe and processing chamber are fabricated from titanium alloy Ti-6Al-4V and are autoclavable. Ease of disassembly facilitates cleaning.
  • Connecting stud: 1/4- 20
  • For low pressure applications only
accessory