Documents
Z549487
Sigma-Aldrich
Stopcock Adapter
straight glass stopcock, joint: ST/NS 19/22
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
straight glass stopcock
Joint
joint: ST/NS19/22
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.