Key Documents
Z547786
Sigma-Aldrich
SafetyBarb® hose connector
straight
Select a Size
1 EA
R$91.00
R$91.00
Estimated to ship onMarch 22, 2025
Select a Size
Change View
1 EA
R$91.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
R$91.00
Estimated to ship onMarch 22, 2025
General description
Consists of PBT cap, PP barbed connector 8.5 mm O.D., and silicone rubber seal. GL14 thread.
Legal Information
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)