Skip to Content
Merck
All Photos(6)

Key Documents

Z545503

Sigma-Aldrich

Molymod® Individual organic model set

DNA (includes stand, 46cm H)

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
R$3,111.00

R$3,111.00


Check Cart for Availability


Select a Size

Change View
1 SET
R$3,111.00

About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

R$3,111.00


Check Cart for Availability

description

base-pairs 10

manufacturer/tradename

Molymod

Looking for similar products? Visit Product Comparison Guide

General description

The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.

Legal Information

Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service