Skip to Content
Merck
All Photos(1)

Documents

Z517399

Sigma-Aldrich

Aldrich® 75 mm O-ring top

Standard taper, top joint: ST/NS 29/32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.43

material

glass

Joint

joint: ST/NS top29/32

sterility

non-sterile

feature

closure type Standard Taper Joint

packaging

pack of 1

manufacturer/tradename

Sigma-Aldrich

Looking for similar products? Visit Product Comparison Guide

General description

Each top comes with a 4 mm PTFE stopcock. Use the stainless steel clamp Z517275.
Torion inlet style − Use 1/4 in. hard-wall transfer tubing (e.g. PP or PTFE) for the airtight transfer of air-sensitive materials.
Standard taper − Joint can accommodate a transfer adapter or a filter funnel.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Torion is a trademark of Bibby Sterlin Ltd.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service