Key Documents
Z509205
Watch glasses
O.D. 40 mm
Select a Size
1 PKG
R$389.00
R$389.00
Check Cart for Availability
Select a Size
Change View
1 PKG
R$389.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
R$389.00
Check Cart for Availability
material
concave
soda-lime glass
sterility
non-sterile
packaging
pkg of 12 ea
manufacturer/tradename
Sigma-Aldrich
O.D.
40 mm
Related Categories
General description
Concave, clear annealed soda lime glass with fire-polished edges.
Choose from one of the most recent versions:
Certificates of Analysis (COA)