Key Documents
Z424595
Pressure relief valve for PLAS-LABS glovebox
Select a Size
1 EA
R$2,900.00
R$2,900.00
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
R$2,900.00
About This Item
UNSPSC Code:
42132201
NACRES:
NB.35
R$2,900.00
Estimated to ship onJune 02, 2025
manufacturer/tradename
Plas-Labs 800-PRV
General description
Useful for eliminating unwanted positive pressure build up in any of Plas-Labs® glove boxes. The acrylic relief valve is easily mounted at the gas inlet or outlet with 1/8 in. NPT SS connections. Exhausted air can be directed away from operators by a hose connection to the outlet port. For minimizing gas consumption, use with the controlled purge system.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)