Skip to Content
Merck
All Photos(1)

Key Documents

Z423122

Sigma-Aldrich

Scienceware® safety labeled wash bottles

Isopropanol

Synonym(s):

lab bottles, safety bottles, wash bottle

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
R$447.00

R$447.00


Check Cart for Availability


Select a Size

Change View
6 EA
R$447.00

About This Item

UNSPSC Code:
41103209
NACRES:
NB.17

R$447.00


Check Cart for Availability

material

low-density polyethylene

feature

with cap

packaging

pack of 6 bottles

manufacturer/tradename

Bel-Art F11646-0624

capacity

500 mL

color

opaque

Looking for similar products? Visit Product Comparison Guide

General description

Safety Labeled wash bottles with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, for making your workplace a safer place.

Features and Benefits

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service