Skip to Content
Merck
All Photos(1)

Key Documents

Z409405

Sigma-Aldrich

Nalgene® safety waste systems

bottle only, capacity 10 L

Synonym(s):

laboratory waste liquid disposal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$1,823.00

R$1,823.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$1,823.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

R$1,823.00


Check Cart for Availability

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 2097-0020

capacity

10 L

Looking for similar products? Visit Product Comparison Guide

General description

Safely dispose of chemical and biological waste and reduce hazardous emissions in the laboratory.

Funnels and waste bottles for the 4L model are also available, sold separately.

Safety waste funnel has a hinged cover (red) to keep emissions contained, funnel attaches to the bottle and remains in place until the bottle is full. A built-in vent minimizes overflow. Bottles are made of safe, durable HDPE.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service