Skip to Content
Merck
All Photos(1)

Key Documents

Z408492

Sigma-Aldrich

Clavies® biohazard autoclave gloves

gauntlet L 5 in. (13 cm), Overall L 13 in. (33 cm)

Synonym(s):

autoclave glove, heat shielding glove, oven glove

Sign Into View Organizational & Contract Pricing

Select a Size

1 PAIR
R$361.00

R$361.00


Check Cart for Availability


Select a Size

Change View
1 PAIR
R$361.00

About This Item

UNSPSC Code:
46181504
NACRES:
HA.03

R$361.00


Check Cart for Availability

material

cotton

size

one size fits all

sterility

non-sterile

feature

CE compliant
latex free
powder-free
reusable

packaging

pkg of 1 pair

manufacturer/tradename

Bel-Art H13201-0000

Overall L

13 in. (33 cm)

gauntlet L

5 in. (13 cm)

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

Soft, pliant, all-cotton terry cloth gloves are excellent for handling hot objects out of an autoclave or oven. Gauntlets available in two lengths and provide wrist and forearm protection.

Features and Benefits

  • Heat-resistant up to 232°C (450°F).
  • Can replace hazardous asbestos gloves.
  • Provides wrist and forearm protection.
  • Bright orange color offers high visibility.
  • Machine wash, air dry

Legal Information

Clavies is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service