Documents
Z265063
Sigma-Aldrich
Butane fuel Micro Torch
MT-51 handheld torch
Synonym(s):
butane torch, butane torches, mini torch
About This Item
UNSPSC Code:
40101834
NACRES:
NB.51
General description
Generate a 2,400°F (1,300°C) pinpoint flame for a wide variety of torch applications. Units are self-igniting and have an adjustable flame. Unit shipped empty, order butane separately.