Skip to Content
Merck
All Photos(2)

Key Documents

Z264385

Supelco

Hamilton® TLC syringes

1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),

coated tip

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$635.00

R$635.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$635.00

About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500
NACRES:
NB.03

R$635.00


Check Cart for Availability

Request a Bulk Order

material

stainless steel needle (PTFE coated tip)

needle size

26s ga

feature

needle type fixed

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80050

needle L

51 mm (2 in.)

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

General description

PTFE coats the final 3/4 in. of this 2 in. (51 mm) long cemented needle. This allows for more reproducible sample spotting on TLC plates.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service