Skip to Content
Merck
All Photos(1)

Key Documents

Z261009

Nalgene® laboratory notebooks

standard paper pages, lined (3 lines per in.)

Synonym(s):

experiment notebook, lab notebook

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$582.00
6 EA
R$2,587.00

R$582.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$582.00
6 EA
R$2,587.00

About This Item

UNSPSC Code:
41100000
NACRES:
AB.01

R$582.00


Check Cart for Availability

publication info

, 96 pp., hard cover

feature

lined (3 lines per in.)

packaging

pack of 1 ea
case of 6 ea

manufacturer/tradename

Nalgene 6301-2000

color

burgundy Cover

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The Nalgene® laboratory notebooks feature durable pages with either a grid or horizontal lines.
  • The pages also contain spaces for dates, signatures of workers, and witnesses for securing patent protection.
  • The book includes pages for instructions on keeping research records, a table of contents, and details of notebook issuing.
  • The polyethylene cover is waterproof.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service