Key Documents
Z247251
Coors porcelain crucible cover
cap size F, I.D. 84 mm
Select a Size
1 PKG
R$1,746.00
R$1,746.00
Check Cart for Availability
Select a Size
Change View
1 PKG
R$1,746.00
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
R$1,746.00
Check Cart for Availability
cap size
F
packaging
pkg of 12 ea
manufacturer/tradename
Coorstek 66127
I.D.
84 mm
General description
Coors porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
  • Ensures great resistance to thermal-shock
  • Durable
  • Rigid
  • Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)