Key Documents
Z243256
ACE Squibb separatory funnel with Locktight stopper
capacity 60 mL, stopper joint: ST/NS 24/40
Select a Size
1 EA
R$1,922.00
R$1,922.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$1,922.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
R$1,922.00
Check Cart for Availability
Joint
joint: ST/NS stopper24/40
manufacturer/tradename
Ace Glass 723155
capacity
60 mL
Related Categories
General description
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)