Key Documents
Z232335
Caps for Duran® laboratory bottles
GL 32 neck thread, blue polypropylene (with lip seal)
Synonym(s):
caps bottles, glass bottles caps
Select a Size
10 EA
R$211.00
R$211.00
Check Cart for Availability
Select a Size
Change View
10 EA
R$211.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$211.00
Check Cart for Availability
material
blue polypropylene (with lip seal)
manufacturer/tradename
DURAN® 292391907
Schott
parameter
-40-140 °C temp. stability
fitting
GL 32 neck thread
General description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)