Skip to Content
Merck
All Photos(1)

Key Documents

Z215996

Solid PTFE stopper

joint: ST/NS 24/29

Synonym(s):

flask stopper, lab stopper, stoppers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$248.00

R$248.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$248.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

R$248.00


Check Cart for Availability

Joint

joint: ST/NS24/29

manufacturer/tradename

Cowie 010.224

Looking for similar products? Visit Product Comparison Guide

General description

Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service