Key Documents
Z171689
Ace double-jacketed coiled condenser
jacket L 250 mm
Select a Size
1 EA
R$3,501.00
R$3,501.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$3,501.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
R$3,501.00
Check Cart for Availability
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 595513
jacket L
250 mm
Related Categories
General description
Extra cooling capacity for volatile materials. Compact design retards diffusion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)