Skip to Content
Merck
All Photos(1)

Key Documents

Z162566

Supelco

Septa for open top caps

for cap size 8 mm, tan PTFE/silicone

Synonym(s):

rubber septum, septum cap, septum caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

tan PTFE/silicone

packaging

pkg of 100

Looking for similar products? Visit Product Comparison Guide

General description

It is used as an air-tight seal, and prevents contamintion
For use with cut-out top caps and Wheaton vials.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service