Z153184
Cochranes molecular models
lattices, minit, classroom set with 5 texts
Synonym(s):
chemistry model sets, molecular model sets
About This Item
Recommended Products
description
Scale: 2 cm = 0.1 nanometer (1 Å), Minit 3 cm = 0.1 nanometer (1 Å), Orbit
feature
no. of atoms 1,470
no. of bonds 1160
manufacturer/tradename
Cochranes 0077
Looking for similar products? Visit Product Comparison Guide
General description
Please see Technical Information Bulletin AL-183 for a listing of these items, available on our website at sigma-aldrich.com
Scale: 2 cm = 0.1 nanometer (1 A), Minit
ATOMS:
- 2-prong linear: Hydrogen, white (125)
- 2-prong 155°: Oxygen, red (30)
- 2-prong 141°: Oxygen, red (30)
- 3-prong 120°: Carbon, black (150), Oxygen, red (150)
- 3-prong 90°, 135°: Oxygen, red (30)
- 4-prong 141°, 116°: Oxygen, red (30)
- 4-prong tetrahedral: Carbon, black (175), Oxygen, red (75), Sulfur, yellow (100), Halogen, green (50), Metal, silver (125)
- 6-prong octahedral: Halogen, green (100), Metal, silver (75)
- 8-prong cubic: Halogen, green (25), Metal, silver (100)
- 12-prong: Metal, silver (100)
BONDS:
- Straw, green, 2.5 cm (640)
- Straw, green, 1.5 cm (320)
- Straw, white, 10 cm (35)
- Straw, white, 3.5 cm (160)
- Straw, mixed set/26 (5)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service