Skip to Content
Merck
All Photos(1)

Documents

Z133205

Whatman® quantitative filter paper, medium speed, Grade 589-WH

Grade 589-WH circles, 110 mm, 100/pk

Synonym(s):

filter papers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

cellulose filters
circles

packaging

pack of 100

manufacturer/tradename

Whatman 10308210
Whatman Article No. 28414152 (US reference)

parameter

Medium flow rate

D

110 mm

pore size

8-10 μm

Looking for similar products? Visit Product Comparison Guide

General description

Ashless filter paper, with high wet strength; hardened (Grade 589-WH)

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service