Key Documents
Z108340
Leather gloves
size L
Select a Size
1 PKG
R$605.00
R$605.00
Check Cart for Availability
Select a Size
Change View
1 PKG
R$605.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
R$605.00
Check Cart for Availability
material
brown gloves
leather
size
L
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 1 pair
manufacturer/tradename
Sigma-Aldrich
General description
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.
Choose from one of the most recent versions:
Certificates of Analysis (COA)