Key Documents
Z104620
PP water aspirator pump
696 mmHg Max. vacuum
Select a Size
1 EA
R$211.00
R$211.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$211.00
About This Item
UNSPSC Code:
41105100
NACRES:
NB.79
R$211.00
Check Cart for Availability
feature
696 mmHg Max. vacuum
manufacturer/tradename
Bel-Art
General description
Operates on water pressure above 11 psi. Includes integral check valve in side arm and 4 3/4 in. length of PE tubing 3/8 in. NPTM faucet connection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)