Skip to Content
Merck
All Photos(3)

Key Documents

X201

Sigma-Aldrich

Xanthene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
Beilstein:
133939
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

bp

310-312 °C (lit.)

mp

101-102 °C (lit.)

SMILES string

C1c2ccccc2Oc3ccccc13

InChI

1S/C13H10O/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-8H,9H2

InChI key

GJCOSYZMQJWQCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthene (10H-9-oxaanthracene) is a heterocyclic compound generally used as an important building block in organic synthesis. It is a core moiety of many dyes like fluorescein, eosins, and rhodamines.[1]

Application

Xanthene can be used as:
  • A reactant in the dehydrogenative cross-coupling reaction with carbonyl compounds.[2][3]
  • A substrate to synthesize N-xanthyl-p-toluenesulfonamide via C−H amination with tosyl azide using rhodium catalyst.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed aerobic asymmetric cross-dehydrogenative coupling of C (sp3)-H bonds driven by visible light
Zhou K, et al.
Green Chemistry, 22(14), 4597-4603 (2020)
Meng Chen et al.
Journal of chromatography. A, 1590, 27-38 (2019-01-12)
An analytical methodology for comprehensive screening of 63 coloring agents of great concern for regulatory control in cosmetics has been established using ultra-high-performance liquid chromatography (UHPLC) coupled with quadrupole-Orbitrap high-resolution mass spectrometry (Q-Orbitrap HRMS). An effective, rapid, and simple sample
Yita Wang et al.
Nanomaterials (Basel, Switzerland), 8(7) (2018-07-07)
The electro-Fenton (EF) process is a type of electrochemical oxidation process; ·OH radicals are generated on the cathode using electricity and decolorize dye wastewaters. Most studies on EF systems in the past have focused on the operating parameters of this
Electrochemical dehydrogenative cross-coupling of xanthenes with ketones
Yang Y-Z, et al.
Chemical Communications (Cambridge, England), 56(14), 7585-7588 (2020)
Nguyen Khoa Hien et al.
ACS omega, 5(33), 21241-21249 (2020-09-03)
A novel coumarin derivative (5) was synthesized and used as a colorimetric and fluorescent probe for selective detection of Cu2+ ions in the presence of other metal ions, with the detection limits of 5.7 and 4.0 ppb, respectively. Cu2+ ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service