Skip to Content
Merck
All Photos(2)

Key Documents

X1376

Sigma-Aldrich

p-Xylene-2-sulfonic acid hydrate

≥98%

Synonym(s):

2,5-Dimethylbenzenesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$953.00

R$953.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
25 G
R$953.00

About This Item

Empirical Formula (Hill Notation):
C8H10O3S · xH2O
CAS Number:
Molecular Weight:
186.23 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$953.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Quality Level

Assay

≥98%

SMILES string

O.Cc1ccc(C)c(c1)S(O)(=O)=O

InChI

1S/C8H10O3S.H2O/c1-6-3-4-7(2)8(5-6)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);1H2

InChI key

VXKQOMCABCHKAQ-UHFFFAOYSA-N

Application

Reagent for serum cholesterol determination.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple method for the determination of serum cholesterol.
D WATSON
Clinica chimica acta; international journal of clinical chemistry, 5, 637-643 (1960-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service