Skip to Content
Merck
All Photos(1)

Documents

WHA9915-2502

Whatman® UNIFLO® 25 syringe filters

polyethersulfone membrane, sterile, pore size 0.2 μm, pack of 200 ea

Synonym(s):

Whatman Syringe Filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

material

polyethersulfone membrane
polypropylene housing

sterility

sterile

packaging

pack of 200 ea

manufacturer/tradename

Whatman 9915-2502

diam.

25 mm

pore size

0.2 μm

fitting

female inlet fitting for Luer Lock
male outlet fitting for Slip Luer

suitability

suitable for (volumes of 100mL or less; filter provides easy separation of solutions)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Whatman® UNIFLO® 25 syringe filters are available with a PES membrane and a polypropylene overmold housing.
  • Chemically resistant PES membrane provides durability, high-temperature resistance, and low protein absorption.
  • Ideally suited for filtering serum, plasma, and tissue culture solutions.
  • Laser-etched printing on the filter facilitates easy identification.
  • Recyclable after use with space-saving packaging.

Features and Benefits

  • 13 or 25 mm diameters
  • 0.2 µm, 0.22 µm or 0.45 µm pore sizes
  • Sterile or non-sterile options
  • Individual printing on the filter for easy identification
  • Bench top space saving packaging

Legal Information

UNIFLO is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service