WHA98949552
Whatman® qualitative filter paper, Grade 597
grade, 597, cellulose filter, filter diam. 55 mm, non-sterile, case of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cellulose filter
sterility
non-sterile
feature
grade 597
packaging
case of 100 ea
manufacturer/tradename
Cytiva 98949552
parameter
70 sec/100 mL speed (Herzberg)
filter diam.
55 mm
thickness
180 μm
pore size
4-7 μm (Particle retention)
basis weight
85 g/m2
Looking for similar products? Visit Product Comparison Guide
General description
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Grade 597: 4-7 μm (medium fast filter paper)
A medium fast filter paper with medium to fine particle retention. Used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).
Grade 597: 4-7 μm (medium fast filter paper)
A medium fast filter paper with medium to fine particle retention. Used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service