Documents
WHA10477602
Whatman® Vacuum filtration apparatus accessories
PZ 001 tweezers, stainless steel
Synonym(s):
Z751979
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
packaging
pk of 1
manufacturer/tradename
Whatman 10477602
Whatman Article No. 28415991 (US reference)
General description
The stainless-steel forceps with smooth angled jaws (104 mm long) are excellent for handling membrane filters.
Features and Benefits
  • They are autoclavable and can be flame sterilized with ethanol.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva