Skip to Content
Merck
All Photos(1)

Key Documents

W522902

Sigma-Aldrich

Musk ketone

≥98%

Synonym(s):

4-tert-Butyl-2,6-dimethyl-3,5-dinitroacetophenone, Ketone Moschus

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6(NO2)2(CH3)2COCH3
CAS Number:
Molecular Weight:
294.30
Beilstein:
2062638
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

grade

Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥98%

composition

Contains EU 1223/2009 restricted Musk Ketone

origin

China origin

mp

135-139 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody

SMILES string

CC(=O)c1c(C)c(c(c(c1C)[N+]([O-])=O)C(C)(C)C)[N+]([O-])=O

InChI

1S/C14H18N2O5/c1-7-10(9(3)17)8(2)13(16(20)21)11(14(4,5)6)12(7)15(18)19/h1-6H3

InChI key

WXCMHFPAUCOJIG-UHFFFAOYSA-N

Other Notes

may contain >0.1% Musk xylene

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>334.4 °F - closed cup

Flash Point(C)

> 168 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence Roosens et al.
Chemosphere, 69(10), 1540-1547 (2007-07-17)
Synthetic musks, such as 7-acetyl-1,1,3,4,4,6-hexamethyl-1,2,3,4-tetrahydronaphthalene (AHTN) and 1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta-gamma-2-benzopyran (HHCB), musk ketone (MK) and musk xylene (MX), are used as an alternative for natural musk. Due to their widespread use, these synthetic compounds turned up in different environmental compartments, such as
Sabine Schnell et al.
Environmental science & technology, 43(24), 9458-9464 (2009-11-26)
Synthetic musks are widely used as perfuming agents in products, such as cosmetics, detergents, and soaps. The increased detection of these substances in the aquatic environment and their high bioconcentration potential raises concerns about potential effects on aquatic species. This
G G Rimkus et al.
Toxicology letters, 111(1-2), 5-15 (2000-01-12)
The monoamino metabolites of the nitro musk fragrances musk xylene (MX) and musk ketone (MK) were analysed simultaneously with their parent compounds by GC/ECD, GC/PND and GC/EI/MS in the various compartments of the aquatic environment. In this review the data
Ana Rita M Silva et al.
Analytical and bioanalytical chemistry, 396(5), 1853-1862 (2010-01-06)
Stir-bar-sorptive extraction with liquid desorption followed by large-volume injection and capillary gas chromatography coupled to mass spectrometry in selected ion monitoring acquisition mode (SBSE-LD/LVI-GC-MS(SIM)) has been developed to monitor ultra-traces of four musks (celestolide (ADBI), galaxolide (HHCB), tonalide (AHTN) and
Yan Lv et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(9), 1125-1130 (2009-09-12)
This study intended to develop a robust and sensitive method for simultaneous determination of polycyclic musks (HHCB and AHTN) and nitro musks (musk xylene (MX) and musk ketone (MK)) in water samples using optimized solid-phase extraction (SPE) by gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service