Skip to Content
Merck
All Photos(1)

Documents

W433600

Sigma-Aldrich

Tridecanoic acid

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CO2H
CAS Number:
Molecular Weight:
214.34
FEMA Number:
4336
Beilstein:
508317
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Assay

≥98%

bp

236 °C/100 mmHg (lit.)

mp

40-44 °C
41-42 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody

SMILES string

CCCCCCCCCCCCC(O)=O

InChI

1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)

InChI key

SZHOJFHSIKHZHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Y Bernard et al.
Pediatric research, 77(6), 829-835 (2015-03-12)
Breastfeeding has been associated with improved cognitive development. This may be explained by polyunsaturated fatty acid (PUFA) content of breast milk, especially long-chain (LC) PUFA that are needed for postnatal brain growth. Using data from the French EDEN cohort, we
S Kargar et al.
Journal of dairy science, 98(7), 4772-4784 (2015-05-20)
Effects of grain type and dietary oil supplement on production performance, energy balance, metabolic heat production, and markers of liver function of heat-loaded lactating dairy cows were evaluated using 8 multiparous Holstein cows (77.0d in milk) in a duplicated 4×4
Xiang Z Li et al.
Animal science journal = Nihon chikusan Gakkaiho, 86(8), 755-764 (2015-01-20)
We hypothesized that manipulating metabolism with fish oil and malate as a hydrogen acceptor would affect the biohydrogenation process of α-linolenic acid by rumen microbes. This study was to examine the effect of fish oil and/or malate on the production
Jacqueline C Lieblein-Boff et al.
PloS one, 10(8), e0136904-e0136904 (2015-09-01)
Lutein is a dietary carotenoid well known for its role as an antioxidant in the macula, and recent reports implicate a role for lutein in cognitive function. Lutein is the dominant carotenoid in both pediatric and geriatric brain tissue. In
S Jorjong et al.
Journal of dairy science, 97(11), 7054-7064 (2014-09-10)
Most cows encounter a state of negative energy balance during the periparturient period, which may lead to metabolic disorders and impaired fertility. The aim of this study was to assess the potential of milk fatty acids as diagnostic tools of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service